APO455838752 Apollo Scientific Ltd

2-(4-Fluorobenzenesulphamido)-2-phenylacetic acid

  • MDL number MFCD00173006
  • Cas Number : 117309-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-181
Synonym N-(4-Fluorobenzenesulphonyl)phenylglycine
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3447

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3447
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart