APO455838753 Apollo Scientific Ltd

2-(4-Fluorobenzenesulphamido)propanoic acid

  • MDL number MFCD00664348
  • Cas Number : 780-97-2
  • Molecular Weight : 247.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 163-165
Iupac Name N-[(4-fluorophenyl)sulfonyl]alanine
Inchi Code 1S/C9H10FNO4S/c1-6(9(12)13)11-16(14,15)8-4-2-7(10)3-5-8/h2-6,11H,1H3,(H,12,13)
Inchi Key JOYNOYIEQOCUEJ-UHFFFAOYSA-N

Description

Synonym N-(4-Fluorobenzenesulphonyl)alanine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 163-165
Product Link https://store.apolloscientific.co.uk/product/2-4-fluorobenzenesulphamidopropanoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-4-fluorobenzenesulphamidopropanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart