APO455838754 Apollo Scientific Ltd

1-(4-Fluorobenzenesulphonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00172808
  • Cas Number : 251096-96-5
  • Molecular Weight : 273.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-99
Synonym N-(4-Fluorobenzenesulphonyl)proline
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-fluorobenzenesulphonylpyrrolidine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-4-fluorobenzenesulphonylpyrrolidine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart