APO455838756 Apollo Scientific Ltd

2,3-Dihydro-5-hydroxy-2-[3-(trifluoromethyl)phenyl]pyridazin-3-one-6-carboxylic acid

  • MDL number MFCD00141002
  • Cas Number : 121582-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 261-263
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3701

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3701
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart