APO455838758 Apollo Scientific Ltd

5-Methoxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD01935988
  • Cas Number : 321848-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-37
Synonym 4-Cyano-5-methoxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3839

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3839
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart