APO455838761 Apollo Scientific Ltd

Methyl 3-(2-chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylate

  • MDL number MFCD00214743
  • Cas Number : 4415-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-55
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3899

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=3899
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart