APO455838767 Apollo Scientific Ltd

Methyl 3-(2-fluoro-4-nitrophenoxy)thiophene-2-carboxylate

  • MDL number MFCD00202715
  • Cas Number : 252026-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-114
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3927

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=3927
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart