APO455838770 Apollo Scientific Ltd

Methyl 1,6-dihydro-4-hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]pyridazin-3-carboxylate

  • MDL number MFCD00141006
  • Cas Number : 121582-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-121
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3936

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3936
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart