APO455838779 Apollo Scientific Ltd

2,3-Difluoro-6-nitroaniline

  • MDL number MFCD00233313
  • Cas Number : 211693-73-1
  • Molecular Weight : 174.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-118
Synonym 2-Amino-3,4-difluoronitrobenzene
Purity 97
Flash Point >110
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-difluoro-6-nitroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluoro-6-nitroaniline
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart