APO455838781 Apollo Scientific Ltd

1-[3-(Trifluoromethyl)benzyl]pyridin-2-one-3-carboxylic acid

  • MDL number MFCD00139983
  • Cas Number : 338754-66-8
  • Molecular Weight : 297.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-161
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-3-trifluoromethylbenzylpyridin-2-one-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-3-trifluoromethylbenzylpyridin-2-one-3-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart