APO455838785 Apollo Scientific Ltd

3-Cyano-5-[(trifluoromethyl)phenyl]-2(1H)-pyridone

  • MDL number MFCD00232370
  • Cas Number : 76053-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234-237
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4244

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=4244
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart