APO455838787 Apollo Scientific Ltd

3-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)propanoic acid

  • MDL number MFCD00173005
  • Cas Number : 250714-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-142
Synonym N-{[3-(Trifluoromethyl)phenyl]sulphonyl}-DL-phenylalanine
Purity 97
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-phenyl-2-3-trifluoromethylphenylsulphonylaminopropanoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-phenyl-2-3-trifluoromethylphenylsulphonylaminopropanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart