APO455838788 Apollo Scientific Ltd

2-[3-(Trifluoromethyl)benzoyl]aminoacetic acid

  • MDL number MFCD00029782
  • Cas Number : 17794-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-138
Synonym N-3-(Trifluoromethyl)benzoylglycine
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5291

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5291
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart