APO455838789 Apollo Scientific Ltd

3-[3-(Trifluoromethyl)benzyloxy]thiophene-2-carboxylic acid

  • MDL number MFCD00173491
  • Cas Number : 339009-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-120
Purity 97
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5313

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5313
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart