APO455838790 Apollo Scientific Ltd

5-[4-(Trifluoromethyl)phenylthio]-1,2,3-thiadiazole-4-carboxylic acid

  • MDL number MFCD01936015
  • Cas Number : 338982-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-166
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5439

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5439
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart