APO455838791 Apollo Scientific Ltd

2-[3-(Trifluoromethyl)benzenesulphonamido]propionic acid

  • MDL number MFCD00172805
  • Cas Number : 288266-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-125
Synonym N-3-(Trifluoromethyl)benzenesulphonylalanine
Purity 97
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-3-trifluoromethylbenzenesulphonamidopropionic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-3-trifluoromethylbenzenesulphonamidopropionic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart