APO455838795 Apollo Scientific Ltd

4-(Difluoromethoxy)benzylhydrazine hydrochloride

  • MDL number MFCD08458142
  • Cas Number : 887596-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10644

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=10644
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart