APO455838796 Apollo Scientific Ltd

Ethyl (3,4-difluorophenyl)(oxo)acetate

  • MDL number MFCD01319619
  • Cas Number : 73790-05-3
  • Molecular Weight : 214.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (3,4-difluorophenyl)(oxo)acetate
Inchi Code 1S/C10H8F2O3/c1-2-15-10(14)9(13)6-3-4-7(11)8(12)5-6/h3-5H,2H2,1H3
Inchi Key DYIBSNLBZKOXNA-UHFFFAOYSA-N

Description

Synonym Ethyl 3,4-difluorophenylglyoxylate, Ethyl 3,4-difluorobenzoylformate
Flash Point 105/0.1mm
Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-34-difluorophenyloxoacetate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-34-difluorophenyloxoacetate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart