APO455838797 Apollo Scientific Ltd

3-Chloro-4-fluorotoluene

  • MDL number MFCD00079730
  • Cas Number : 1513-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-1-fluoro-4-methylbenzene
Purity 97
Density 1.18
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 167-169
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-fluorotoluene

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-fluorotoluene
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart