APO455838799 Apollo Scientific Ltd

2,2-Dichloro-3,3,3-trifluoropropan-1-ol

  • MDL number MFCD04039302
  • Cas Number : 20411-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2-Dichloro-3-hydroxy-1,1,1-trifluoropropane, 2,2-Dichloro-2-(trifluoromethyl)ethan-1-ol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6447

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6447
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart