APO455838800 Apollo Scientific Ltd

2-Fluoro-5-hydroxybenzoic acid

  • MDL number MFCD01862084
  • Cas Number : 51446-30-1
  • Molecular Weight : 156.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-206
Synonym 3-Carboxy-4-fluorophenol
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-hydroxybenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-hydroxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart