APO455838803 Apollo Scientific Ltd

1-Fluoro-3-(trifluoromethoxy)benzene

  • MDL number MFCD00236323
  • Cas Number : 1077-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.33
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 100-105
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5200

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5200
pictograms GHS02;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart