APO455838805 Apollo Scientific Ltd

4-Bromo-2-fluoro-6-nitroanisole

  • MDL number MFCD00013374
  • Cas Number : 74266-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-56
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10618

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=10618
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart