APO455838808 Apollo Scientific Ltd

3-Chloro-4-fluorobenzyl chloride

  • MDL number MFCD03701044
  • Cas Number : 2994-69-6
  • Molecular Weight : 179.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-4-(chloromethyl)-1-fluorobenzene, alpha,3-Dichloro-4-fluorotoluene
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-fluorobenzyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-fluorobenzyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart