APO455838811 Apollo Scientific Ltd

Ethyl 2,4,5-trifluorobenzoylacetate

  • MDL number MFCD00792431
  • Cas Number : 98349-24-7
  • Molecular Weight : 246.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-64
Synonym Ethyl 3-oxo-3-(2,4,5-trifluorophenyl)propanoate
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 92/0.08mm
Product Link https://store.apolloscientific.co.uk/product/ethyl-245-trifluorobenzoylacetate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-245-trifluorobenzoylacetate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart