APO455838815 Apollo Scientific Ltd

3-(Trifluoromethoxy)phenylacetylene

  • MDL number MFCD08458148
  • Cas Number : 866683-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 23 - 55
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point >35
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9543

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=9543
pictograms GHS02;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart