APO455838820 Apollo Scientific Ltd

4-Bromo-3-fluorobenzonitrile

  • MDL number MFCD07782069
  • Cas Number : 133059-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9549

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=9549
Availability details for
Quantity
  • Availability
  • Add to Cart