APO455838823 Apollo Scientific Ltd

3-Bromo-4-methylbenzotrifluoride

  • MDL number MFCD01631584
  • Cas Number : 66417-30-9
  • Molecular Weight : 239.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-4-(trifluoromethyl)toluene
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 88
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-methylbenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-methylbenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart