APO455838827 Apollo Scientific Ltd

2-Amino-5-fluorobenzeneboronic acid, neopentyl glycol ester, N-BOC protected

  • MDL number MFCD07776961
  • Cas Number : 1218791-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[(tert-Butoxycarbonyl)amino]-5-fluorobenzeneboronic acid, neopentyl glycol ester, tert-Butyl [2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-4-fluorophenyl]carbamate
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9558

Safety Information

signalword Warning
pcodes P280;P302+P352;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9558
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart