APO455838832 Apollo Scientific Ltd

2,2-Dichloro-1,1-difluoroethyl methyl ether

  • MDL number MFCD00040144
  • Cas Number : 76-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -35
Density 1.426
Flash Point 37
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 103-104.5
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8258

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=8258
pictograms GHS02;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart