APO455838850 Apollo Scientific Ltd

4,4,5,5,5-Pentafluoropentanoic acid

  • MDL number MFCD08458152
  • Cas Number : 3637-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 23-25
Synonym 4,4,5,5,5-Pentafluorovaleric acid
Flash Point >110
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 84/20mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10617

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10617
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart