APO455838851 Apollo Scientific Ltd

2-(2-Fluorophenyl)amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide

  • MDL number MFCD08458153
  • Cas Number : 1211414-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225-228
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10616

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=10616
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart