APO455838858 Apollo Scientific Ltd

2-Chloro-3-fluorobenzonitrile

  • MDL number MFCD03407966
  • Cas Number : 874781-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-56
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10611

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=10611
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart