APO455838859 Apollo Scientific Ltd

2-Chloro-3-fluorobromobenzene

  • MDL number MFCD04038306
  • Cas Number : 883499-24-9
  • Molecular Weight : 209.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29
Synonym 1-Bromo-2-chloro-3-fluorobenzene
Density 1.70
Flash Point 78
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-3-fluorobromobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-3-fluorobromobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart