APO455838861 Apollo Scientific Ltd

6-Fluoro-1H-indole-3-carboxaldehyde

  • MDL number MFCD00069703
  • Cas Number : 2795-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-176
Synonym 6-Fluoro-3-formyl-1H-indole
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-fluoro-1h-indole-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-fluoro-1h-indole-3-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart