APO455838862 Apollo Scientific Ltd

1-(4-Fluorophenyl)pyrrolidin-2-one

  • MDL number MFCD00138504
  • Cas Number : 54660-08-1
  • Molecular Weight : 179.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-57
Synonym 1-(4-Fluorophenyl)-2-oxopyrrolidine, 1-Fluoro-4-(2-oxopyrrolidin-1-yl)benzene
Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-fluorophenylpyrrolidin-2-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-4-fluorophenylpyrrolidin-2-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart