APO455838869 Apollo Scientific Ltd

2-(Trifluoromethoxy)phenol

  • MDL number MFCD00236324
  • Cas Number : 32858-93-8
  • Molecular Weight : 178.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 82
Iupac Name 2-(trifluoromethoxy)phenol
density 1.332
Inchi Code 1S/C7H5F3O2/c8-7(9,10)12-6-4-2-1-3-5(6)11/h1-4,11H
Inchi Key GQWMNVOVQZIPJC-UHFFFAOYSA-N

Description

Melting Point 82
Synonym 2-Hydroxyphenyl trifluoromethyl ether
Purity 98%
Density 1.332
Flash Point 69-71/60mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethoxyphenol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethoxyphenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart