APO455838875 Apollo Scientific Ltd

4-Amino-3-fluorobenzoic acid

  • MDL number MFCD01660374
  • Cas Number : 455-87-8
  • Molecular Weight : 155.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-218
Synonym 4-Carboxy-2-fluoroaniline
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-3-fluorobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-3-fluorobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart