APO455838877 Apollo Scientific Ltd

1H-Perfluoropentane

  • MDL number MFCD08458172
  • Cas Number : 375-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.63
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 42
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10566

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=10566
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart