APO455838880 Apollo Scientific Ltd

Perfluoro-2,5,8,11-tetraoxadodecane

  • MDL number MFCD08458174
  • Cas Number : 64028-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Perfluorotriglyme, Perfluoro(triethylene glycol dimethyl ether), 1,1,1,3,3,4,4,6,6,7,7,9,9,10,10,12,12,12-Octadecafluoro-2,5,8,11-tetraoxadodecane
Purity 98
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 106
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10564

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10564
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart