APO455838881 Apollo Scientific Ltd

Perfluorodiglyme

  • MDL number MFCD08458175
  • Cas Number : 40891-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Perfluoro(diethylene glycol dimethyl ether)
Purity 98
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 66
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10563

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10563
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart