APO455838886 Apollo Scientific Ltd

4-Hydroxy-2-(pyrazin-2-yl)-6-(trifluoromethyl)pyrimidine

  • MDL number MFCD00665787
  • Cas Number : 438249-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3704

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=3704
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart