APO455838888 Apollo Scientific Ltd

3,5-Bis(trifluoromethyl)-alpha-methylbenzylamine

  • MDL number MFCD04038875
  • Cas Number : 187085-97-8
  • Molecular Weight : 257.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine
Purity 97
Flash Point 66
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 6.5-64.5/3.2mm
Product Link https://store.apolloscientific.co.uk/product/35-bistrifluoromethyl-alpha-methylbenzylamine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-bistrifluoromethyl-alpha-methylbenzylamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart