APO455838894 Apollo Scientific Ltd

Methyl 2-amino-5-fluorobenzoate

  • MDL number MFCD03791308
  • Cas Number : 319-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Fluoro-2-(methoxycarbonyl)aniline, Methyl 5-fluoroanthranilate
Storage temp. Store at 2-8
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9523

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=9523
Availability details for
Quantity
  • Availability
  • Add to Cart