APO455838897 Apollo Scientific Ltd

2-Amino-3-fluorobenzonitrile

  • MDL number MFCD07368774
  • Cas Number : 115661-37-5
  • Molecular Weight : 136.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-80
Synonym 2-Cyano-6-fluoroaniline, 3-Fluoroanthranilonitrile
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-3-fluorobenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-3-fluorobenzonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart