APO455838908 Apollo Scientific Ltd

Difluoro(phenyl)acetonitrile

  • MDL number MFCD07784286
  • Cas Number : 2002-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2-Difluoro-2-phenylethanenitrile
Purity 97
Storage temp. Store at 2-8
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10542

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=10542
Availability details for
Quantity
  • Availability
  • Add to Cart