APO455838911 Apollo Scientific Ltd

4-(Trifluoromethyl)-1H-imidazole

  • MDL number MFCD08458860
  • Cas Number : 33468-69-8
  • Molecular Weight : 136.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-151
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethyl-1h-imidazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethyl-1h-imidazole
pictograms GHS07;GHS08
hstatements H302;H315;H319;H335;H351
Availability details for
Quantity
  • Availability
  • Add to Cart