APO455838912 Apollo Scientific Ltd

1H-Tridecafluoro-3,3-dimethylhex-1-yne

  • MDL number MFCD00155807
  • Cas Number : 261503-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.56
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 89
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3509

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=3509
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart