APO455838913 Apollo Scientific Ltd

2-Chloro-4-iodo-5-(trifluoromethyl)pyridine

  • MDL number MFCD01862099
  • Cas Number : 505084-55-9
  • Molecular Weight : 307.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-127
Synonym 6-Chloro-4-iodo-alpha,alpha,alpha-trifluoro-3-picoline
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-iodo-5-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-iodo-5-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart