APO455838919 Apollo Scientific Ltd

2-Methyl-4-(trifluoromethoxy)acetanilide

  • MDL number MFCD07368678
  • Cas Number : 886762-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9191

Safety Information

signalword Warning
pcodes P280;P271;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9191
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart